Learn More
Medchemexpress LLC HY-110287 5mg Medchemexpress, Apcin CAS:300815-04-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1102875MG
Encompass_Preferred
Medchemexpress, HY-110287 5mg Apcin CAS:300815-04-7 Apcin, a ligand of Cdc20, is a potent and competitive anaphase-promoting complex/cyclosome (APC/C(Cdc20)) E3 ligase activity inhibitor. Apcin competitively inhibits APC/C-dependent ubiquitylation by binding to Cdc20 and preventing substrate recognition. Apcin occupes the D-box-binding pocket on the side face of the WD40-domain and can prolong mitosis[2][3]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9685