Learn More
AdipoGen Apcin (5 mg)
SDP
Supplier:  AdipoGen AGCR13603M005
Encompass_Preferred
Chemical. CAS: 300815-04-7. Formula: C13H14Cl3N7O4. MW: 438.7. Cell permeable compound capable of slowing anaphase-promoting complex/cyclosome (APC/C)-dependent proteolysis. Inhibitor of the ubiquitin ligase activity of the APC/C by inhibiting substrate binding to Cdc20. Distinct mechanism of action than proTAME. Blocks mitotic exit only modestly when used alone in cultured cells. A strong synergistic effect is observed when Apcin and proTAME are used simultaneously, providing a more robust mitotic arrest. This may be very beneficial when treating cells that do not efficiently convert proTAME to its active parent compound (TAME) or if experiments demand strong APC/C inhibition.
Catalog No. 50-205-3971