Learn More
Medchemexpress LLC Apcin-a | 1683617-62-0 | 98.0% | C10H14Cl3N5O2 | 10MG
SDP
Catalog No. 5000260987
Encompass_Preferred
Apcin-A is a small-molecule inhibitor that selectively targets the cell division cycle protein Cdc20. It competitively binds the D-box pocket of Cdc20 to inhibit APC/C-Cdc20-mediated substrate ubiquitination, blocking binding of proteins such as securin and cyclin B1 and interfering with anaphase initiation. Supplied as a research reagent for cellular and biochemical studies.
  • Selective Cdc20 inhibition for mitotic regulation studies.
  • Derivative of apcin that blocks substrate ubiquitination.
  • High purity (98.0%) for reproducible results.
  • Soluble in DMSO at 100 mg/mL for convenient stock solutions.
  • Solid, off-white to yellow appearance; stable under recommended storage.
  • Intended for research use only; not for human or clinical use.
Catalog No. 50-002-60987 Supplier Medchemexpress LLC Supplier No. HY13084110MG
Apcin-A is a small-molecule inhibitor that selectively targets the cell division cycle protein Cdc20. It competitively binds the D-box pocket of Cdc20 to inhibit APC/C-Cdc20-mediated substrate ubiquitination, blocking binding of proteins such as securin and cyclin B1 and interfering with anaphase initiation. Supplied as a research reagent for cellular and biochemical studies.
  • Selective Cdc20 inhibition for mitotic regulation studies.
  • Derivative of apcin that blocks substrate ubiquitination.
  • High purity (98.0%) for reproducible results.
  • Soluble in DMSO at 100 mg/mL for convenient stock solutions.
  • Solid, off-white to yellow appearance; stable under recommended storage.
  • Intended for research use only; not for human or clinical use.