Learn More
Bellco Glass, Inc. APD Kit (OLD)-5 Position For 7630 - Rear Mount Units
SDP
Catalog No. 501946072
Encompass_Preferred
This option is used for converting: 7730-75000,7730-75500 7730-76000, 7730-76500, 7730-75220, 7730-75520, 7730-76220 & 7730-76520 five position roller apparatus to all positions drive. This option is not required for use with glass bottles but is desirable for use with plastic disposable roller bottles.. MADE IN THE USA
Catalog No. 50-194-6072 Supplier Bellco Glass, Inc. Supplier No. 773050555
This option is used for converting: 7730-75000,7730-75500 7730-76000, 7730-76500, 7730-75220, 7730-75520, 7730-76220 & 7730-76520 five position roller apparatus to all positions drive. This option is not required for use with glass bottles but is desirable for use with plastic disposable roller bottles.. MADE IN THE USA