Learn More
Bellco Glass, Inc. APD Retrofit Kit, 5 Position For 7630 Series
SDP
Catalog No. 501945904
Encompass_Preferred
This option is used for converting: 7630-75000,7630-75012 7630-00509, 7630-00510, 7630-75220, 7630-75222, 7630-22509 & 7630-22510 five Position ition roller apparatus to all Position itions drive. ALL Position ITION DRIVE CONVERSION KITADAPTS ANY REAR DRIVE ROLLER APPARATUS UNIT TO OPERATE WITH PLASTIC BOTTLES(1) APD needed for *each* deck of Roller Apparatus. MADE IN THE USA
Catalog No. 50-194-5904 Supplier Bellco Glass, Inc. Supplier No. 763050555
This option is used for converting: 7630-75000,7630-75012 7630-00509, 7630-00510, 7630-75220, 7630-75222, 7630-22509 & 7630-22510 five Position ition roller apparatus to all Position itions drive. ALL Position ITION DRIVE CONVERSION KITADAPTS ANY REAR DRIVE ROLLER APPARATUS UNIT TO OPERATE WITH PLASTIC BOTTLES(1) APD needed for *each* deck of Roller Apparatus. MADE IN THE USA