Learn More
Medchemexpress LLC Ape1-in-2 | 2923433-95-6 | 98.3% | 522.21 g/mol | C9H12Cl2N4O5Pt | 10 MG
SDP
Supplier:  Medchemexpress LLC HY15188310MG
Encompass_Preferred
APE1-IN-2 is a platinum(IV) proagent that inhibits apurinic/apyrimidinic endonuclease 1 (APE1). It has demonstrated anticancer activity in preclinical studies by promoting intracellular platinum accumulation and activating DNA damage response and apoptosis pathways. Supplied as a solid research compound, it is intended for laboratory use only.
Catalog No. 50-002-85892