Learn More
Aviva Systems Biology Corp APEH Antibody - middle region (ARP48182_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48182_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against APEH. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human APEH. Alternate names: ACPH, APH, D3F15S2, D3S48E, DNF15S2, MGC2178, OPH, AAREApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-285-349