Learn More
Aviva Systems Biology Corp APEH Antibody - N-terminal region (ARP48181_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48181_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against APEH. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human APEH. Alternate names: ACPH, APH, D3F15S2, D3S48E, DNF15S2, MGC2178, OPH, AAREApplicationWB, IHC. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-54037