Learn More
New England Biolabs, Inc. ApeKI – 1250 units
SDP
Catalog No. 50812397
Encompass
A restriction endonuclease that recognizes the sequence G^CWG_C.
Catalog No. 50-812-397 Supplier New England Biolabs, Inc. Supplier No. R0643L
A restriction endonuclease that recognizes the sequence G^CWG_C.