Learn More
CPC Scientific Glp-Arg-Pro-Arg-Leu-Ser-His-Lys-Gly-Pro-Met-Pro-Phe-OH (trifluoroacetate salt) 0.5MG
SDP
Supplier:  CPC Scientific APEL002A
Encompass
Pyroglutamyl form of apelin-13 that displays a high activity with an EC50 value of 0.30 nM.SEQUENCE: Glp-Arg-Pro-Arg-Leu-Ser-His-Lys-Gly-Pro-Met-Pro-Phe-OH (trifluoroacetate salt)ONE-LETTER SEQUENCE: Glp-RPRLSHKGPMPFMOLECULAR FORMULA: C69H108N21O16S1MOLECULAR WEIGHT: 1533.81STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [217082-60-5]SYNONYMS: (Pyr)-Apelin-13 (human, bovine, mouse, rat)RESEARCH AREA: AntimicrobialREFERENCES:K. Tatemoto et al., Biochem Biophy. Res. Commun., 251, 471 (1998)SKU(s): APEL-002A, APEL-002B
Catalog No. 50-210-8780