Learn More
Eurofins Discoverx Apelin-13 TFA salt, 1 mg. (92-1093)
Supplier:  Eurofins Discoverx 921093
Encompass
Apelin-13 is a bioactive neuropeptide processed from the pre-proprotein apelin that is active in the nervous system. It is an endogenous ligand of the receptor APJ (EC50=0.37 nM). Plays a role in cardiovascular regulation, fluid homeostasis, hypertension, and insulin sensitivity. May undergo pyroglutamylation, and may block HIV-1 and HIV-2 entry into cells expressing the APJ receptor. Compared to apelin-36, blocks HIV entry poorly. Provided as 1 mg of dry powder alongside reconstitution buffer. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.
Catalog No. 50-199-9631