Learn More
Medchemexpress LLC Apelin-16, human, bovine acetate | 99.8% | 2010.37 (free base) | 5 MG
SDP
Catalog No. 5000398910
Encompass_Preferred
Apelin-16, human, bovine acetate is a biologically active peptide. Apelin is the endogenous ligand for the G-protein-coupled APJ receptor. It is produced as a pre-proprotein of 77 amino acids, and Apelin-16 is one of its active fragments.
  • Biologically active peptide
  • Endogenous ligand for the G-protein-coupled APJ receptor
  • Derived from a pre-proprotein of 77 amino acids
  • Active fragment of Apelin
Catalog No. 50-003-98910 Supplier Medchemexpress LLC Supplier No. HYP5392A5MG
Apelin-16, human, bovine acetate is a biologically active peptide. Apelin is the endogenous ligand for the G-protein-coupled APJ receptor. It is produced as a pre-proprotein of 77 amino acids, and Apelin-16 is one of its active fragments.
  • Biologically active peptide
  • Endogenous ligand for the G-protein-coupled APJ receptor
  • Derived from a pre-proprotein of 77 amino acids
  • Active fragment of Apelin