Learn More
Abcam Apelin 17, Endogenous apelin receptor (APJ) agonist, 1MG
Catalog No. 502706973
Encompass_Preferred
MW 2138.6 Da, Purity >95%. Apelin 17, Endogenous apelin receptor (APJ) agonist. Achieve your results faster with highly validated, pure and trusted compounds.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6973 Supplier Abcam Supplier No. AB1410111MG
MW 2138.6 Da, Purity >95%. Apelin 17, Endogenous apelin receptor (APJ) agonist. Achieve your results faster with highly validated, pure and trusted compounds.
The product is subject to the following: Abcam Restricted Use Statement