Learn More
CPC Scientific H-Leu-Val-Gln-Pro-Arg-Gly-Ser-Arg-Asn-Gly-Pro-Gly-Pro-Trp-Gln-Gly-Gly-Arg-Arg-Lys-Phe-Arg-Arg-Gln-Arg-Pro-Arg-Leu-Ser-His-Lys-Gly-Pro-Met-Pro-Phe-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific APEL001B
Encompass
Orphan G protein-coupled receptor shown to be a coreceptor for simian and human immunodeficiency virus (SIV and HIV) strains. As long as apelin is an endogenous ligand for the APJ receptor, inhibitory effects of apelin peptides on HIV infection have been found to display that apelin peptides inhibit the entry of some HIV-1 and HIV-2 strains into NP-2/CD4 cells expressing APJ. Strong inhibitory efficiency.ONE-LETTER SEQUENCE: LVQPRGSRNGPGPWQGGRRKFRRQRPRLSHKGPMPFMOLECULAR FORMULA: C184H297N69O43S1MOLECULAR WEIGHT:4195.89STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [252642-12-9], SYNONYMS: Apelin-36 (human)RESEARCH AREA: Antimicrobial
Catalog No. 50-210-8779