Learn More
Abcam Apelin 36 (human), Endogenous apelin receptor (APJ) agonist, 1MG
Catalog No. 502706975
Encompass_Preferred
MW 4195.87 g/mol. Endogenous apelin receptor (APJ) agonist (EC₅₀ = 20 nM). Exhibits cardioprotective effect and inhibits entry of HIV-1 and HIV-2 into NP-2/CD4 cells. Active in vivo.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6975 Supplier Abcam Supplier No. AB1410091MG
MW 4195.87 g/mol. Endogenous apelin receptor (APJ) agonist (EC₅₀ = 20 nM). Exhibits cardioprotective effect and inhibits entry of HIV-1 and HIV-2 into NP-2/CD4 cells. Active in vivo.
The product is subject to the following: Abcam Restricted Use Statement