Learn More
Cayman Chemical ApelIn-36trIfluoroACTAte500ug
SDP
Catalog No. 502955820
Encompass_Preferred
Full-length mature apelin peptide; acts primarily in the regulation of cardiovascular function, fluid homeostasis, hypertension, and insulin sensitivity; a less potent APJ agonist than either apelin-17 or apelin-13 (EC50 = 20, 2.5, and 0.37 nM, respectively); potently inhibits HIV-1 entry into cells expressing APJ and CD4
Catalog No. 50-295-5820 Supplier Cayman Chemical Supplier No. 13524500 UG
Full-length mature apelin peptide; acts primarily in the regulation of cardiovascular function, fluid homeostasis, hypertension, and insulin sensitivity; a less potent APJ agonist than either apelin-17 or apelin-13 (EC50 = 20, 2.5, and 0.37 nM, respectively); potently inhibits HIV-1 entry into cells expressing APJ and CD4