Learn More
Electron Microscopy Sciences Willco-dish Glass Bottom Kit Assembly Device 50x20 mm, 22 mm Glass Aperture
SDP
Catalog No. 501900296
Encompass_Preferred
Round, anodized, aluminum “Assembly Devices" for a quick, accurate and safe positioning and assembly of the glass coverslip and the adhesive ring. 50x20 mm.
Aperture: 22 mm
Catalog No. 50-190-0296 Supplier Electron Microscopy Sciences Supplier No. 7066521
Round, anodized, aluminum “Assembly Devices" for a quick, accurate and safe positioning and assembly of the glass coverslip and the adhesive ring. 50x20 mm.
Aperture: 22 mm