Learn More
Waters Corp Waters Quality Parts Aperture Disc, 0.2 mm, Stainless Steel, 5 pk
Catalog No. 502224486 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These replacement aperture discs are designed for use in the source of ACQUITY RDa and BioAccord Systems.
  • Product type: Spare parts kits
  • Units per package: 5 pk
  • Type: Mechanical
  • System type: MS
  • Material: Stainless steel
  • Internal diameter: 0.2 mm
  • Replacement schedule: As required
Catalog No. 50-222-4486 Supplier Waters Corp Supplier No. 700012106
These replacement aperture discs are designed for use in the source of ACQUITY RDa and BioAccord Systems.
  • Product type: Spare parts kits
  • Units per package: 5 pk
  • Type: Mechanical
  • System type: MS
  • Material: Stainless steel
  • Internal diameter: 0.2 mm
  • Replacement schedule: As required