Learn More
Electron Microscopy Sciences Gilder Aperture Grid with Single 1500 µm hole, Nickel
SDP
Catalog No. 5028643
Encompass_Preferred
Single Slot aperture grids have a thickness of are 50 microns. Aperture diameter is 1500 um.
Material: Nickel
Catalog No. 50-286-43 Supplier Electron Microscopy Sciences Supplier No. GA1500NI
Single Slot aperture grids have a thickness of are 50 microns. Aperture diameter is 1500 um.
Material: Nickel