Learn More
Electron Microscopy Sciences Gilder Aperture Grid with Single 2000 µm hole, Nickel
SDP
Catalog No. 5028647
Encompass_Preferred
Single Slot aperture grids have a thickness of are 50 microns. Aperture diameter is 2000 um.
Material: Nickel
Catalog No. 50-286-47 Supplier Electron Microscopy Sciences Supplier No. GA2000NI
Single Slot aperture grids have a thickness of are 50 microns. Aperture diameter is 2000 um.
Material: Nickel