Learn More
Electron Microscopy Sciences Gilder Aperture Grid with Single 600 µm hole, Copper
SDP
Catalog No. 5028639
Encompass_Preferred
Single Slot aperture grids have a thickness of are 50 microns. Aperture diameter is 600 um.
Material: Copper
Catalog No. 50-286-39 Supplier Electron Microscopy Sciences Supplier No. GA600CU
Single Slot aperture grids have a thickness of are 50 microns. Aperture diameter is 600 um.
Material: Copper