Learn More
Electron Microscopy Sciences Gilder Aperture Grid with Single 75 µm hole, Copper
SDP
Catalog No. 5028637
Encompass_Preferred
Single Slot aperture grids have a thickness of are 50 microns. Aperture diameter is 75 um.
Material: Copper
Catalog No. 50-286-37 Supplier Electron Microscopy Sciences Supplier No. GA75CU
Single Slot aperture grids have a thickness of are 50 microns. Aperture diameter is 75 um.
Material: Copper