Learn More
Electron Microscopy Sciences Gilder Aperture Grid with Single 800 µm hole, Copper
SDP
Catalog No. 5028635
Encompass_Preferred
Single Slot aperture grids have a thickness of are 50 microns. Aperture diameter is 800 um.
Material: Copper
Catalog No. 50-286-35 Supplier Electron Microscopy Sciences Supplier No. GA800CU
Single Slot aperture grids have a thickness of are 50 microns. Aperture diameter is 800 um.
Material: Copper