Learn More
Electron Microscopy Sciences Gilder Aperture Grid with Single 800 µm hole, Nickel
SDP
Catalog No. 5028636
Encompass_Preferred
Single Slot aperture grids have a thickness of are 50 microns. Aperture diameter is 800 um.
Material: Nickel
Catalog No. 50-286-36 Supplier Electron Microscopy Sciences Supplier No. GA800NI
Single Slot aperture grids have a thickness of are 50 microns. Aperture diameter is 800 um.
Material: Nickel