Learn More
Electron Microscopy Sciences EMS Molybdenum Aperture with 100 µm hole, 6.35 mm O.D., 0.006 mm thick
SDP
Catalog No. 501901602
Encompass_Preferred
Molybdenum disc aperture with 100 um hole for use with ETEC electron microscopes.
O.D.: 6.35 mm
Thickness: 0.006 mm
Catalog No. 50-190-1602 Supplier Electron Microscopy Sciences Supplier No. 47100MO
Molybdenum disc aperture with 100 um hole for use with ETEC electron microscopes.
O.D.: 6.35 mm
Thickness: 0.006 mm