Learn More
Electron Microscopy Sciences EMS Molybdenum Aperture with 10 µm hole, 2 mm O.D., 0.53 mm thick
SDP
Catalog No. 501901314
Encompass_Preferred
Molybdenum disc aperture with 10 um hole for use with Cambridge, Siemens, JEOL, and Cameca electron microscopes.
O.D.: 2 mm
Thickness: 0.53 mm
Catalog No. 50-190-1314 Supplier Electron Microscopy Sciences Supplier No. 42010MO
Molybdenum disc aperture with 10 um hole for use with Cambridge, Siemens, JEOL, and Cameca electron microscopes.
O.D.: 2 mm
Thickness: 0.53 mm