Learn More
Electron Microscopy Sciences EMS Molybdenum Aperture with 30 µm hole, 2 mm O.D., 0.24 mm thick
SDP
Catalog No. 501901309
Encompass_Preferred
Molybdenum disc aperture with 30 um hole for use with Phillips and Cameca electron microscopes.
O.D.: 2 mm
Thickness: 0.24 mm
Catalog No. 50-190-1309 Supplier Electron Microscopy Sciences Supplier No. 40030MO
Molybdenum disc aperture with 30 um hole for use with Phillips and Cameca electron microscopes.
O.D.: 2 mm
Thickness: 0.24 mm