Learn More
Electron Microscopy Sciences EMS Molybdenum Aperture with 40 µm hole, 2 mm O.D., 0.53 mm thick
SDP
Catalog No. 501901318
Encompass_Preferred
Molybdenum disc aperture with 40 um hole for use with Cambridge, Siemens, JEOL, and Cameca electron microscopes.
O.D.: 2 mm
Thickness: 0.53 mm
Catalog No. 50-190-1318 Supplier Electron Microscopy Sciences Supplier No. 42040MO
Molybdenum disc aperture with 40 um hole for use with Cambridge, Siemens, JEOL, and Cameca electron microscopes.
O.D.: 2 mm
Thickness: 0.53 mm