Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Electron Microscopy Sciences EMS Molybdenum Aperture with 50 µm hole, 3 mm O.D., 0.165 mm thick
SDP

Catalog No. 501901592
Encompass_Preferred


O.D.: 3 mm
Thickness: 0.165 mm

Catalog No. 50-190-1592 Supplier Electron Microscopy Sciences Supplier No. 45050MO
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'


O.D.: 3 mm
Thickness: 0.165 mm

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.