Learn More
Abcam APET x 2, ASIC3 channel blocker, 100UG
Catalog No. 502706409
Encompass_Preferred
MW 4561.05 g/mol, Purity >98%. Selective, reversible ASIC3 channel blocker (IC₅₀ = 63 nM). Inhibits heteromeric ASIC2b and ASIC3 currents (IC₅₀ = 117 nM) and NaV1.8 channels (IC₅₀ = 2.6 μM). Shows analgesic effects in vivo.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6409 Supplier Abcam Supplier No. AB141849100UG
MW 4561.05 g/mol, Purity >98%. Selective, reversible ASIC3 channel blocker (IC₅₀ = 63 nM). Inhibits heteromeric ASIC2b and ASIC3 currents (IC₅₀ = 117 nM) and NaV1.8 channels (IC₅₀ = 2.6 μM). Shows analgesic effects in vivo.
The product is subject to the following: Abcam Restricted Use Statement