Learn More
Mac-Mod Avantor Apex ODS, 100 A, 5 um, 4.6 x 250 mm HPLC Column
SDP
Supplier:  Mac-Mod 4M25310
Encompass
Avantor Apex and Avantor Apex ll phases are recommended for routine analyses. Conventional 100 pore size spherical silica particle column. Narrow particle size distribution.
Catalog No. 50-112-0513