Learn More
Perkin Elmer US LLC Apex HF system for NexION
Catalog No. 502112652
Encompass_Preferred
Apex HF system for the PE NexION 300. Includes one PFA-100 ST nebulizer and adapter to NexION.
Catalog No. 50-211-2652 Supplier Perkin Elmer US LLC Supplier No. N8145292
Add to Cart
Add to Cart
Request a Quote
Apex HF system for the PE NexION 300. Includes one PFA-100 ST nebulizer and adapter to NexION.