Learn More
Genesee Scientific Corporation Apex Taq Red Master Mix, 2X Concentrate, 1.5 mM MgCl2, Direct Gel Loading, 1000 Reactions
SDP
Catalog No. NC3346002
Encompass
The Apex Taq RED Master Mix, 2X, is a ready-to-use solution for efficient DNA amplification via PCR. It simplifies setup by combining polymerase, buffer, ultra-pure dNTPs, and red dye into one concentrate, minimizing contamination risk. This product allows direct loading of PCR products on agarose gels, enhancing visualization. With high product yield and the ability to process amplicons up to 5 kb, it stands out for its time-saving convenience and user-friendly design.
Catalog No. NC3346002 Supplier Genesee Scientific Corporation Supplier No. 42138B
The Apex Taq RED Master Mix, 2X, is a ready-to-use solution for efficient DNA amplification via PCR. It simplifies setup by combining polymerase, buffer, ultra-pure dNTPs, and red dye into one concentrate, minimizing contamination risk. This product allows direct loading of PCR products on agarose gels, enhancing visualization. With high product yield and the ability to process amplicons up to 5 kb, it stands out for its time-saving convenience and user-friendly design.