Learn More
Aviva Systems Biology Corp APEX1 Antibody - N-terminal region (ARP32651_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32651_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against APEX1. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human APEX1. Alternate names: APE, APX, APE1, APEN, APEX, HAP1, REF1ApplicationIHC, WB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-54692