Learn More
Aviva Systems Biology Corp APEX1 Antibody - N-terminal region (ARP37013_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37013_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against APEX1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: APE, Apex, HAP1, Ref-1ApplicationWB. Species reactivity: Cow, Guinea Pig, Horse, Mouse, Rabbit, Rat
Catalog No. 50-014-56311