Learn More
Aviva Systems Biology Corp APEX1 Antibody - N-terminal region (ARP37014_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37014_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against APEX1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of mouse APEX1. Alternate names: APE, Apex, HAP1, Ref-1ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-56312