Learn More
Aviva Systems Biology Corp APEX1 Antibody - N-terminal region (ARP58027_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58027_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against APEX1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human APEX1. Alternate names: APE, APE1, APEN, APEX, APX, HAP1, REF-1, REF1ApplicationWB. Species reactivity: Human, Pig
Catalog No. 50-286-428