Learn More
Apexbio Technology LLC ApexPrep DNA Plasmid Miniprep Kit 300 tests
SDP
Catalog No. 5000061775
Encompass_Preferred
The ApexPrep DNA Plasmid Miniprep Kit is designed for isolation of plasmid DNA through alkaline-mediated bacterial cell lysis Upon lysis plasmid DNA selectively binds to a silica-based spin column under high-salt conditions separating it from cellular debris and contaminants Subsequent washing removes proteins and organic impurities yielding purified plasmid DNA (up to 30 g) from bacterial culture volumes of 1 5 mL Extracted plasmid DNA is suitable for downstream molecular biology applications including restriction digestion PCR amplification sequencing cloning transfection and in vitro transcription or translation assays
Catalog No. 50-000-61775 Supplier Apexbio Technology LLC Supplier No. A5001300
The ApexPrep DNA Plasmid Miniprep Kit is designed for isolation of plasmid DNA through alkaline-mediated bacterial cell lysis Upon lysis plasmid DNA selectively binds to a silica-based spin column under high-salt conditions separating it from cellular debris and contaminants Subsequent washing removes proteins and organic impurities yielding purified plasmid DNA (up to 30 g) from bacterial culture volumes of 1 5 mL Extracted plasmid DNA is suitable for downstream molecular biology applications including restriction digestion PCR amplification sequencing cloning transfection and in vitro transcription or translation assays