Learn More
MSC APF KIT
Catalog No. 5000677223
Encompass
The 3M™ Speedglas™ G5-01 Heavy-Duty Welding Helmet 1000 APF Kit delivers advanced respiratory protection. By integrating a large head cover and fabric shroud, this kit achieves a NIOSH/CSA Z94.4 Assigned Protection Factor (APF) of 1000 when used with compatible powered air purifying respirators (PAPRs), enhancing safety in demanding environments.
  • Achieves APF 1000 with compatible PAPRs for enhanced respiratory protection.
  • Integrates seamlessly with Adflo™ and Versaflo™ PAPR and Supplied Air Systems.
  • Features an innovative G5-01 helmet duct system for customizable airflow.
  • Includes an extra-large comfort headrest and self-adjusting headband for improved comfort.
  • Designed for heavy-duty, high-amperage welding and frequent grinding applications.
Catalog No. 50-006-77223 Supplier MSC Supplier No. 18481440
The 3M™ Speedglas™ G5-01 Heavy-Duty Welding Helmet 1000 APF Kit delivers advanced respiratory protection. By integrating a large head cover and fabric shroud, this kit achieves a NIOSH/CSA Z94.4 Assigned Protection Factor (APF) of 1000 when used with compatible powered air purifying respirators (PAPRs), enhancing safety in demanding environments.
  • Achieves APF 1000 with compatible PAPRs for enhanced respiratory protection.
  • Integrates seamlessly with Adflo™ and Versaflo™ PAPR and Supplied Air Systems.
  • Features an innovative G5-01 helmet duct system for customizable airflow.
  • Includes an extra-large comfort headrest and self-adjusting headband for improved comfort.
  • Designed for heavy-duty, high-amperage welding and frequent grinding applications.