Learn More
Medchemexpress LLC Apg-2449 | 2196186-84-0 | 98.9% | 640.24 g/mol | C33H42ClN5O4S | 10 MG
SDP
Catalog No. 5000286366
Encompass_Preferred
APG-2449 is an orally active small-molecule inhibitor that targets focal adhesion kinase (FAK) and receptor tyrosine kinases including ALK and ROS1. It is provided for research use only and is characterized by defined physicochemical properties and supporting analytical documentation.
  • Inhibits FAK, ALK, and ROS1 kinases.
  • Demonstrates oral activity in preclinical models.
  • High purity suitable for research applications.
  • Supplied in small-scale packaging for dosing and formulation studies.
  • Characterized by molecular formula C33H42ClN5O4S and molecular weight 640.24 g/mol.
Catalog No. 50-002-86366 Supplier Medchemexpress LLC Supplier No. HY15516310MG
APG-2449 is an orally active small-molecule inhibitor that targets focal adhesion kinase (FAK) and receptor tyrosine kinases including ALK and ROS1. It is provided for research use only and is characterized by defined physicochemical properties and supporting analytical documentation.
  • Inhibits FAK, ALK, and ROS1 kinases.
  • Demonstrates oral activity in preclinical models.
  • High purity suitable for research applications.
  • Supplied in small-scale packaging for dosing and formulation studies.
  • Characterized by molecular formula C33H42ClN5O4S and molecular weight 640.24 g/mol.