Learn More
Selleck Chemical LLC APG-2449-E1508-25MG
SDP
Catalog No. 503134544
Encompass_Preferred
APG-2449 is an orally active inhibitor of ALK/ROS1/FAK APG-2449 exerts potent and durable antitumor activity in human non-small-cell lung cancer (NSCLC) and ovarian tumor models
Catalog No. 50-313-4544 Supplier Selleck Chemical LLC Supplier No. E150825MG
APG-2449 is an orally active inhibitor of ALK/ROS1/FAK APG-2449 exerts potent and durable antitumor activity in human non-small-cell lung cancer (NSCLC) and ovarian tumor models