Learn More
Aviva Systems Biology Corp APG4B Antibody - C-terminal region (ARP34434_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34434_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against APG4B. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human APG4B. Alternate names: APG4B, AUTL1ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-52956