Learn More
Waters Corp APGC Corona Discharge Pin Assembly
Catalog No. 50793052 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Replacement Corona Pin Assembly. Used in conjunction with APGC source as an alternative form of ion generation. Common to both APGC and APGC 2.
Catalog No. 50-793-052 Supplier Waters Corp Supplier No. 700004809
Replacement Corona Pin Assembly. Used in conjunction with APGC source as an alternative form of ion generation. Common to both APGC and APGC 2.