Learn More
Waters Corp Waters APGC Transfer Line Locking Ring, Replacement
Catalog No. 50793054 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The APGC Transfer Line Locking Ring is a replacement component designed for the APGC Heated Transfer Line.
  • Provides secure locking for the APGC Heated Transfer Line.
  • Compatible with MS system types.
  • Used with instruments including MS Sources, Xevo TQ-XS, and APGC.
Catalog No. 50-793-054 Supplier Waters Corp Supplier No. 700004815
The APGC Transfer Line Locking Ring is a replacement component designed for the APGC Heated Transfer Line.
  • Provides secure locking for the APGC Heated Transfer Line.
  • Compatible with MS system types.
  • Used with instruments including MS Sources, Xevo TQ-XS, and APGC.