Learn More
Waters Corp WATERS CORPORATION Active Pre-Heater, Stainless Steel, 18.5 in.
Catalog No. 501462019 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This product is a Replacement Active Preheater (APH) made of Stainless Steel, 18.5 in. long. It is utilized within ACQUITY CM-A and ACQUITY APC CM-S systems.
  • Replacement part
  • Designed for CM-A and APC CM-S systems
  • Stainless steel construction
Catalog No. 50-146-2019 Supplier Waters Corp Supplier No. 205000774
This product is a Replacement Active Preheater (APH) made of Stainless Steel, 18.5 in. long. It is utilized within ACQUITY CM-A and ACQUITY APC CM-S systems.
  • Replacement part
  • Designed for CM-A and APC CM-S systems
  • Stainless steel construction