Learn More
Waters Corp Active PreHeater (APH), SS, .004 in ID x 23.5 in, with Gasket - PLUS
Catalog No. NC1648517 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Replacement Active PreHeater (APH), SS, .004 in ID x 23.5 in, with Gasket - PLUS. ; This APH is used in H-Class PLUS and I-Class PLUS systems with CH-30A.
Catalog No. NC1648517 Supplier Waters Corp Supplier No. 205001759
Replacement Active PreHeater (APH), SS, .004 in ID x 23.5 in, with Gasket - PLUS. ; This APH is used in H-Class PLUS and I-Class PLUS systems with CH-30A.