Learn More
ABclonal Technology APH1A Rabbit pAb
SDP
Supplier:  ABclonal Technology A14666500UL
Encompass_Preferred
This gene encodes a component of the gamma secretase complex that cleaves integral membrane proteins such as Notch receptors and beta-amyloid precursor protein. The gamma secretase complex contains this gene product, or the paralogous anterior pharynx defective 1 homolog B (APH1B), along with the presenilin, nicastrin, and presenilin enhancer-2 proteins. The precise function of this seven-transmembrane-domain protein is unknown though it is suspected of facilitating the association of nicastrin and presenilin in the gamma secretase complex as well as interacting with substrates of the gamma secretase complex prior to their proteolytic processing. Polymorphisms in a promoter region of this gene have been associated with an increased risk for developing sporadic Alzheimers disease. Alternative splicing results in multiple protein-coding and non-protein-coding transcript variants.
Catalog No. 50-001-04639