Learn More
Innovative Research Inc Pooled Human Plasma Apheresis Derived Na Heparin 100ml
Supplier:  Innovative Research Inc IPLANAH100ML
Encompass_Preferred
Pooled Human Plasma from Innovative Research is apheresis derived. The plasma is collected from donors in an FDA-approved collection center. The plasma is collected via apheresis, aliquoted to customer specifications, and frozen. 
Please note that this product does not allow for donor requests off-the-shelf, but we can create custom pooled lots to your exact specifications upon request. Contact us for details & pricing. 
Viral Testing
Each unit is tested for the standard FDA-required viral markers, and found negative for HBsAg, HCV, HIV-1, HIV-2, HIV-1Ag or HIV-1 NAT, ALT, and syphilis using FDA-approved methods. 
Anticoagulants
We are able to add to this Na Citrate-collected product an anticoagulant that will work best for your research. Our standard anticoagulant options include K2 EDTA, K3 EDTA, Na EDTA, Na Citrate, Na Heparin, and Li Heparin,
Catalog No. 50-203-6344