Learn More
AG Scientific Inc Aphidicolin, 1 MG
Catalog No. 502236877
Encompass_Preferred
Aphidicolin is a reversible inhibitor of nuclear DNA replication in eukaryotes and some animal viruses. It interferes with dCTP-specific binding sites on DNA polymerase-alpha. It also works on other DNA polymerases, however it does not work on DNA polymerase-alpha. Viruses inducing DNA polymerase-alpha, such as Herpes simplex, can be affected by aphidicolin because of its mechanism. It could be used for clinical study. Isolated from Nigrospora oryzae.CAS Number: 38966-21-1Molecular Weight: 338.5Chemical Formula: C20H34O4Solubility: Methanol: Clear colorless solution at 10 mg/mlStorage Temperature: 2-8CResearch or further manufacturing use only, not for food or drug use.
Catalog No. 50-223-6877 Supplier AG Scientific Inc Supplier No. A10261MG
Aphidicolin is a reversible inhibitor of nuclear DNA replication in eukaryotes and some animal viruses. It interferes with dCTP-specific binding sites on DNA polymerase-alpha. It also works on other DNA polymerases, however it does not work on DNA polymerase-alpha. Viruses inducing DNA polymerase-alpha, such as Herpes simplex, can be affected by aphidicolin because of its mechanism. It could be used for clinical study. Isolated from Nigrospora oryzae.CAS Number: 38966-21-1Molecular Weight: 338.5Chemical Formula: C20H34O4Solubility: Methanol: Clear colorless solution at 10 mg/mlStorage Temperature: 2-8CResearch or further manufacturing use only, not for food or drug use.