Learn More
AdipoGen (+)-Aphidicolin,Chemical. CAS: 38966-21-1
SDP
Catalog No. 50464948
Encompass_Preferred
(+)-Aphidicolin, AdipoGen Life Sciences. Chemical. CAS: 38966-21-1. Formula: C20H34O4. MW: 338.5. Isolated from Phoma sp. BS 7210. Phytotoxin. Antibiotic. Antiviral and antineoplastic agent. Antileishmanial agent. Reversible inhibitor of eukaryotic nuclear DNA replication. Useful for cell synchronization. Blocks the cell cycle at G1/S phase. Prolongs the half life of DNA methyltransferase. DNA methylation/demethylation modulator. Specific DNA polymerase alpha and delta inhibitor in eukaryotic cells and in some viruses of animal origin. Acts synergistically with vincristine and doxorubicin. Apoptosis inhibitor/inducer.
Catalog No. 50-464-948 Supplier AdipoGen Supplier No. BVT0307M001
(+)-Aphidicolin, AdipoGen Life Sciences. Chemical. CAS: 38966-21-1. Formula: C20H34O4. MW: 338.5. Isolated from Phoma sp. BS 7210. Phytotoxin. Antibiotic. Antiviral and antineoplastic agent. Antileishmanial agent. Reversible inhibitor of eukaryotic nuclear DNA replication. Useful for cell synchronization. Blocks the cell cycle at G1/S phase. Prolongs the half life of DNA methyltransferase. DNA methylation/demethylation modulator. Specific DNA polymerase alpha and delta inhibitor in eukaryotic cells and in some viruses of animal origin. Acts synergistically with vincristine and doxorubicin. Apoptosis inhibitor/inducer.